Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

66886-U

Supelco

Empore SPE Disks

SDB-RPS, diam. 47 mm, pk of 20

Synonym(s):

polymeric reverse phase w/Strong Cation Exchange, SPE disk, 47 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NB.21

product name

Empore SPE Disks, matrix active group SDB-RPS, diam. 47 mm, pk of 20

description

polystyrene-divinylbenzene, reversed-phase sulfonate

Quality Level

product line

Empore

packaging

pk of 20

manufacturer/tradename

CDS Analytical 98-0604-0226-4

technique(s)

solid phase extraction (SPE): suitable

diam.

47 mm

membrane thickness

0.5 mm

surface area

450 m2/g

matrix active group

SDB-RPS

particle size

12 μm (spherical)
12 μm

pore size

80 Å pore size

Looking for similar products? Visit Product Comparison Guide

General description

Empore membrane SPE technology comprises of SPE particles tightly enmeshed within a network of inert PTFE fibrils (90% SPE particles : 10% PTFE, by weight). The SPE-membrane fabrication process results in a highly dense and uniform extraction medium that offers distinct advantages over traditional sorbent/packed-bed SPE products. Empore SPE technology provides a denser, more uniform extraction bed than traditional packed bed products allowing for smaller bed weights, shorter analyte to pore diffusion paths, and more efficient extractions.

The Empore line of SPE disks comprises of the most complete line of SPE disks for extracting large volumes of aqueous samples. The product line ranges from time-tested C18 to unique phase chemistries such as carbon and the highly polar Oil & Grease disk. The disks are ideal for environmental analysis where 1 L sample volumes are not uncommon and provide an efficient alternative to LLE.

Save Time & Money:
  • Reduced SPE Bed Mass = Reduced SPE Solvent & Elution Volumes
  • Minimize SPE eluate evaporation time
  • Potentially allows for direct injection of the SPE eluate
  • Dense & Uniform Extraction Medium = No SPE Channeling and Voiding
  • Efficient mass-transfer kinetics allow for faster flow rates
  • Eliminate SPE fines improving column and instrument life

Application


  • Determination of diphenylarsinic acid, phenylarsonic acid and inorganic arsenic in drinking water: This study utilizes Empore SPE Disks for the simultaneous separation and preconcentration of toxic compounds in drinking water, highlighting the disks′ application in environmental safety and public health (Hagiwara et al., 2013).

  • Monitoring of ecotoxicological hazard potential by polar organic micropollutants: The research emphasizes the utility of SPE disks in the environmental monitoring of micropollutants, contributing significantly to the assessment of water quality and ecological risks (Escher et al., 2008).

  • Determination of commonly used polar herbicides in agricultural drainage waters: Demonstrates the efficiency of SPE Disks in analyzing herbicide residues in agricultural runoff, providing insights into pollution control and chemical residue management (Tran et al., 2007).

  • Stability of pesticides on solid-phase extraction disks: Explores the stability of various pesticides on SPE disks under different conditions, proving the disks′ reliability for sample storage and analysis in multi-environmental studies (Cobb et al., 2006).

Other Notes

Developed for the mixed-mode extraction of polar and non-polar analytes from water samples and soil extracts. Offers unique selectivity for analytes that exhibit polar functional groups. Because the phase contains sulfonic acid functional groups, secondary SCX interactions can occur towards basic compounds that contain amine functional groups.

  • Explosive residues (HDX, RDX)

Legal Information

Empore is a trademark of CDS Analytical

Disclaimer

CDS Analytical LLC recently acquired the rights to Empore products from 3M Purification Inc. For an interim phase, you may receive products packaged with either manfacturer′s branding.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immanuel D Green et al.
Nucleic acids research, 48(12), 6513-6529 (2020-05-26)
Monocytes and macrophages are essential components of the innate immune system. Herein, we report that intron retention (IR) plays an important role in the development and function of these cells. Using Illumina mRNA sequencing, Nanopore direct cDNA sequencing and proteomics
Alexandra K Davies et al.
Nature communications, 9(1), 3958-3958 (2018-09-29)
Adaptor protein 4 (AP-4) is an ancient membrane trafficking complex, whose function has largely remained elusive. In humans, AP-4 deficiency causes a severe neurological disorder of unknown aetiology. We apply unbiased proteomic methods, including 'Dynamic Organellar Maps', to find proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service