Key Documents
57627
Sample Loop for Model 7725 Injector
volume 100 μL
About This Item
UNSPSC Code:
41115854
volume
100 μL
General description
Sample loop for model 7725 injector (volume 100 μL) is best suited for advanced fluid-handling solutions for broad range of analytical instrumentation and in vitro diagnostic systems. It is useful in high performance liquid chromatographic (HPLC) systems and is quite accurate and precise in injecting 1 μL-5 mL concentration of samples.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Articles
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.