Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55061

Mobile Phase Reservoir System

bottle size 2 L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

bottle size

2 L

Looking for similar products? Visit Product Comparison Guide

General description

This convenient system is made entirely of inert parts: glass, Tefzel, PEEK, and PTFE TFE. Includes 10 ft./3 m of 1/8 in. O.D. PTFE tubing, a PEEK bottom-of-the-bottle filter/sparger, a plug, and a 1-liter or 2-liter safety-coated bottle.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

replacement part

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service