Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54373

Supelco

Pyridalyl

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2-{3-{2,6-Dichloro-4-[(3,3-dichloro-2-propen-1-yl)oxy]phenoxy}propoxy}-5-(trifluoromethyl)pyridine, [2,6-Dichloro-4-(3,3-dichloroallyloxy)phenyl]-3-(5-trifluoromethyl-2-pyridyloxy)propyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14Cl4F3NO3
CAS Number:
Molecular Weight:
491.12
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

−20°C

SMILES string

ClC1=C(OCCCOC2=NC=C(C(F)(F)F)C=C2)C(Cl)=CC(OCC=C(Cl)Cl)=C1

InChI

1S/C18H14Cl4F3NO3/c19-13-8-12(27-7-4-15(21)22)9-14(20)17(13)29-6-1-5-28-16-3-2-11(10-26-16)18(23,24)25/h2-4,8-10H,1,5-7H2

InChI key

AEHJMNVBLRLZKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service