Skip to Content
MilliporeSigma
All Photos(1)

Documents

504920

Supelco

Supelclean LC-SCX SPE Tube

bed wt. 100 mg, volume 1 mL, pk of 108

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712

material

polypropylene tube

feature

endcapped

composition

bed wt., 100 mg

packaging

pk of 108

technique(s)

solid phase extraction (SPE): suitable

volume

1 mL

matrix active group

SCX phase
SCX

diameter

40-45 μm

pore size

60 Å

application(s)

food and beverages

separation technique

ion exchange
ion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Supelclean LC-SCX SPE Tubes consist of sulfonic acid bonded silica with Na+ counterions. They can be used for the strong cation exchange of cations, antibiotics, drugs, organic bases, amino acids, catecholamines, herbicides, nucleic acids and bases, nucleosides, and surfactants.

Application

Supelclean LC-SCX SPE Tube may be used for the solid-phase extraction of β-agonist compounds in feed samples and adulterants in slimming functional foods prior to their quantification by chromatographic techniques.

Legal Information

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A GC-EI-MS-MS Method for Simultaneous Determination of Seven Adulterants in Slimming Functional Foods.
Li Y, et al.
Journal of Chromatographic Science, 50(10), 928-933 (2012)
Simultaneous determination of cimaterol, salbutamol, terbutaline and ractopamine in feed by SPE coupled to UPLC.
Qu CH, et al.
Chromatographia, 73(3-4), 243-249 (2011)

Articles

SPE retention mechanism in this case is based on the electrostatic attraction of charged functional groups of the analyte(s) to oppositely charged functional groups on the sorbent.

Protocols

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service