Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48613

Supelco

1,2–Dichloroethane solution

certified reference material, 200 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4Cl2
CAS Number:
Molecular Weight:
98.96
EC Number:
MDL number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

ClCCCl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2

InChI key

WSLDOOZREJYCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2−Dichloroethane is a chlorinated hydrocarbon, which is produced in highest volumes compared to any other chlorinated derivatives of hydrocarbons.

Application

1,2−Dichloroethane may be used as a reference standard for the determination of the analyte in water by automated purge and trap (PT) coupled to gas chromatography-mass spectrometry (GC-MS), and in pharmaceutical products by headspace-programmed temperature vaporization-fast GC-MS and in municipal wastewater by headspace−solid phase microextraction−gas chromatography (HS-SPME-GC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of chlorinated volatile organic compounds in water and municipal wastewater using headspace -solid phase microextraction -gas chromatography
Antoniou CV, et al.
Journal of Chromatography A, 1132(1-2), 310 -3314 (2006)
Analysis of class 1 residual solvents in pharmaceuticals using headspace-programmed temperature vaporization-fast gas chromatography -mass spectrometry
Pavon JLP, et al.
Journal of Chromatography A, 1141(1), 123 -1130 (2007)
Evidence of Substantial Carbon Isotope Fractionation among Substrate, Inorganic Carbon, and Biomass during Aerobic Mineralization of 1, 2-Dichloroethane byXanthobacter autotrophicus
Hunkeler D and Aravena R
Applied and Environmental Microbiology, 66(11), 4870 -44876 (2000)
Multicomponent analysis of volatile organic compounds in water by automated purge and trap coupled to gas chromatography -mass spectrometry
Martinez E, et al.
Journal of Chromatography A, 959(1-2), 181 -1190 (2002)
Yoko Imai et al.
The journal of physical chemistry. B, 116(1), 585-592 (2011-12-15)
The electron transfer (ET) between cytochrome c (Cyt c) in water (W) and 1,1'-dimethylferrocene (DiMFc) in 1,2-dichloroethane (DCE) was studied. The cyclic voltammograms obtained for the interfacial ET under various conditions could be well reproduced by digital simulation based on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service