Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48566

Supelco

Acenaphthylene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8
CAS Number:
Molecular Weight:
152.19
Beilstein:
774092
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

280 °C (lit.)

mp

78-82 °C (lit.)

density

0.899 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

c1cc2C=Cc3cccc(c1)c23

InChI

1S/C12H8/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-8H

InChI key

HXGDTGSAIMULJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acenaphthylene may be used as an analytical reference standard for the determination of the analyte in water using  gas chromatography-flame ionization detection (GC-FID).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

251.6 °F - closed cup

Flash Point(C)

122.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of organic compounds in water using dispersive liquid-liquid microextraction.
Rezaee M, et al.
Journal of Chromatography A, 1116(1-2), 1-9 (2006)
E Leonhardt et al.
Analytical chemistry, 70(6), 1228-1230 (1998-04-08)
Polycyclic aromatic hydrocarbons were extracted from a soil sample using ultrasound and dichloromethane-, cyclohexane-, and toluene-water mixtures. It was found that when dichloromethane is used as an extractant, acenaphthylene reacts with the solvent. Several chlorinated and oxygenated derivatives were identified.
Yan Hao et al.
Organic & biomolecular chemistry, 10(4), 724-728 (2011-12-15)
A simple and novel protocol for the efficient synthesis of a series of 8-carboxylnaphthyl functionalized pyrazolo[3,4-b]pyridine derivatives was developed through a one-pot, three-component reaction involving acenaphthylene-1,2-dione and 1H-pyrazol-5-amine in acetic acid medium. The reaction represents the first facile conversion of
K Schirmer et al.
Toxicology, 127(1-3), 129-141 (1998-08-12)
Sixteen polycyclic aromatic hydrocarbons (PAHs) were screened for their ability to be directly cytotoxic to a cell line from the rainbow trout gill, RTgill-W1. Exposure times of 2 h or less were sufficient for direct cytotoxicity to be detected, which
Ming Chen et al.
Chemical communications (Cambridge, England), (9)(9), 1112-1114 (2008-02-23)
A higher efficiency of excitation energy transfer occurs to a luminescent diphenylanthracenyl acceptor incorporated at the centre, rather than the end, of an acenaphthylene polymer chain.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service