Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

26887-U

SGE Syringes for Agilent/HP 7673, 7683 and 6850 ALS Autosampler, Fixed Needle

volume 5 μL, 5F-HP-0.63/0.47, needle size 23-26 ga, needle L 42 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

cone tip stainless steel needle (point style 1)

needle size

23-26 ga

feature

needle type fixed tapered needle

packaging

pkg of 6 ea

manufacturer/tradename

SGE 001822

needle L

42 mm

volume

5 μL

Looking for similar products? Visit Product Comparison Guide

General description

SGE syringes are designed and tested to meet critical autosampler specifications. These syringes are designed with a unique plunger to eliminate bending and seizing. The needle point style withstands multiple and fast septum injections, making it one of the most troublefree autosampler syringes available.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service