Skip to Content
MilliporeSigma
All Photos(11)

Key Documents

ZRB5054

Sigma-Aldrich

Anti-Calretinin Antibody, clone 3K22, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3K22, monoclonal
recombinant monoclonal

description

3K22 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 31.4 kDa
observed mol wt ~30 kDa

species reactivity

mouse, rat, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable using 1:100
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:400
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 3K22 is a ZooMAb rabbit recombinant monoclonal antibody that detects Calretinin. It targets an epitope with in the internal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 99 amino acids from the internal region of rat Calretinin.

Application

Anti-Calretinin, clone 3K22, ZooMAb, Cat. No. ZRB5054, is a recombinant rabbit monoclonal antibody that detects Calretinin and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1000 dilution from a representative lot detected Calretinin in rat and human brain tissue lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Calretinin in PC12 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:400 dilution from a representative lot detected Calretinin in human cerebral cortex tissue sections.

Target description

Calretinin (UniProt: P47728; also known as CR) is encoded by the Calb2 gene (Gene ID: 117059) in rat. Calretinin belongs to the calbindin protein family and acts as a calcium-binding protein. It is expressed in both central and peripheral nervous system and is found abundantly in auditory neurons. It is involved in regulating cytosolic calcium ion concentrations and has six calcium-binding domains known as EF-hands. Its expression is shown to be regulated at the level of transcription by nuclear respiratory factor 1 (NRF-1) and E2F2 transcription factors. In rat striatum calretinin neurons form a distinct subset of medium-sized (7- 20 μm), aspiny, GABAergic interneurons. Calretinin can also serve as an important differential diagnostic tool for cystic and solid ameloblastic tumors. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Petryszyn, S., et al. (2017). J. Neural Transm. doi: 10.1007/s00702-017-1687; Kresoja-Rakic, J., et al. (2017). Front. Genet. doi: 10.3389/fgene.2017.00070).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

120 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service