Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

ZRB2726

Sigma-Aldrich

Anti-LDH-A Antibody, clone 1I7 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Cell proliferation-inducing gene 19 protein, EC 1.1.1.27, L-lactate dehydrogenase A chain, LDH muscle subunit′LDH-M, Renal carcinoma antigen NY-REN-59

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 1I7
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 36.69 kDa
observed mol wt ~37 kDa

purified by

using Protein A

species reactivity

mouse, monkey, human, rat

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... LDHA(3939)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1I7 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects L-lactate dehydrogenase A (LDH-A). It targets an epitope within 15 amino acids within the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human L-lactate dehydrogenase A.

Application

Quality Control Testing

Evaluated by Western Blotting in NIH3T3 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected LDH-A in NIH3T3 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected LDH-A in lysates from COS-7, K562, and PC12 cells.

Affinity Binding Assay: A representative lot of this antibody bound LDH-A peptide with a KD of 3.2 x 10-6 in an affinity binding assay.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected LDH-A in one million K562 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected LDH-A in Human liver tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

L-Lactate dehydrogenase A chain (EC 1.1.1.27; UniProt P00338; also known as Cell proliferation-inducing gene 19 protein, LDH muscle subunit, LDH-A, LDH-M, Renal carcinoma antigen NY-REN-59) chain (EC 1.1.1.27; UniProt P00338; also known as Cell proliferation-inducing gene 19 protein, LDH muscle subunit, LDH-A, LDH-M, Renal carcinoma antigen NY-REN-59) is encoded by the LDHA (also known as GSD11, PIG19) gene (Gene ID 3939) in human. LDH is a homotetrameric, cytosolic enzyme that is found at high levels in tissues that utilize glucose for energy. It catalyzes the reversible conversion of pyruvate to lactate with the regeneration of NADH to NAD+. This reaction involves the transfer of a hydride ion from NADH to the C2 carbon of pyruvate. This conversion is essential in hypoxic and anaerobic conditions when ATP production by oxidative phosphorylation is disrupted. Detection of LDH in serum indicates organ and tissue damage and it serves as a good marker for screening of tissue injuries. Five tetrameric isozymes of LDH have been described that are composed of two different types of subunits (M-muscle type and H-heart type), which can yield five combinations: H4; H3M; H2M2; HM3; and M4. LDH-A is the muscle specific isoform is expressed mainly in the skeletal muscle. A sequence of ten amino acids in its N-terminal region is reported to be important for its structural stability, enzyme activity, and sensitivity to denaturing environments. Cancer cells are known to use LDH-A to increase the rate of glycolysis and production of ATP and lactate even in the presence of oxygen. This switch to anaerobic glycolysis benefits cancer cells by avoiding generation of oxidative stress by the electron transport chain. Overexpression of LDH-A is associated with several poor prognostic factors, including tumor hypoxia, angiogenesis, proliferation, and resistance to chemo- and radiotherapy. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonal. (Ref.: Valvona, CJ., et al. (2016). Brain Pathol. 26(1); 3-17; Kim, JW., and Dang, CV. (2006). Cancer Res. 66(18); 8927-8930).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service