Skip to Content
MilliporeSigma
All Photos(12)

Key Documents

ZRB1930

Sigma-Aldrich

Anti-SEH1L Antibody, clone 1E11 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Nucleoporin SEH1;GATOR complex protein SEH1;Nup107-160 subcomplex subunit SEH1;SEC13-like protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E11, recombinant monoclonal

description

1E11 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 39.65 kDa
observed mol wt ~35 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

rat, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... SEH1L(81929)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E11 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Nucleoporin SEH1L. It targets an epitope within 21 amino acids from the C-terminal region.

Immunogen

KLH-conjugate linear peptide corresponding to 21 amino acids from the C-terminal region of human Nucleoporin SEH1, isoform A.

Application

Quality Control Testing

Evaluated by Western Blotting in PC3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SEH1L in PC3 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected SEH1L in MDA-MB-231 and A549 cell lysates.

Affinity Binding Assay: A representative lot of this antibody bound SEH1L with a KD of 2.4 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected SEH1L in A549 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected SEH1L in human cerebral cortex tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in PC3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SEH1L in PC3 cell lysate.

Target description

Nucleoporin SEH1 (UniProt: Q96EE3; also known as GATOR complex protein SEH1, Nup107-160 subcomplex subunit SEH1, SEC13-like protein, SEH1L) is encoded by the SEH1L (also known as SEC13L, SEH1) gene (Gene ID: 81929) in human. Nuclear pore complexes (NPCs) are composed of multiple copies of approximately 30 different proteins termed as nucleoporins. In vertebrates, one such building block of the NPC, the Nup107-160 complex, contains six different nucleoporins. SEH1L is one of the components of the Nup107-160 subcomplex. This subcomplex is required for normal kinetochore microtubule attachment, mitotic progression, and chromosome segregation. The SEH1L subunit that contains six WD domains appears to be only weakly associated with the Nup107-160 subcomplex. Depletion of Nup107-160 complex or depletion of SEH1L is shown to induce mitotic delay. SEH1L depleted cells also display impaired chromosome congression, reduced kinetochore tension, defects in and kinetochore microtubule attachments. SEH1L is also a component of the GATOR subcomplex 2 and functions within the amino acid-sensing branch of TORC1 signaling pathway. It is reported to indirectly activate mTORC1 signaling pathway through the inhibition of the GATOR 1 subcomplex. Knockdown of SEH1L is reported to inhibit mTORC1 activity. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Parmigiani, A., et al. (2014). Cell Rep. 9(4); 1281-1291; Zuccolo, M., et al. (2007). EMBO J. 26(7); 1853-1864; Loiodice, I., et al. (2004). Mol. Biol. Cell. 15(7); 3333-3344).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service