Skip to Content
MilliporeSigma
All Photos(15)

Key Documents

ZRB1654

Sigma-Aldrich

Anti-CXCL10 Antibody, clone 3F14 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

C-X-C motif chemokine 10;10 kDa interferon gamma-induced protein;Gamma-IP10;IP-10;Small-inducible cytokine B10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3F14, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 10.88 kDa
observed mol wt ~10 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CXCL10(3627)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3F14 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects C-X-C motif chemokine 10 (CXCL10). It targets an epitope within 16 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal half of human C-X-C motif chemokine 10 (CXCL10).

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant Human CXCL10.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected recombinant Human CXCL10.

Tested Applications

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected CXCL10 in human thymus and human tonsil tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CXCL10 in human thymus tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound CXCL10 peptide with a KD of 1.3 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

C-X-C motif chemokine 10 (UniProt: P02778; also known as 10 kDa interferon gamma-induced protein, Gamma-IP10, IP-10, Small-inducible cytokine B10) is encoded by the CXCL10 (also known as INP10, SCYB10) gene (Gene ID: 3627) in human. CXCL10 is a secreted protein that is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off to produce the mature form. It is mainly secreted by monocytes, endothelial cells as well as fibroblasts and is expressed by epithelial cells in thymus tissue. Microglial cells are also reported to produce CXCL10 in response to viral stimulation. It is a pro-inflammatory cytokine that is involved in a wide variety of processes, including chemotaxis, differentiation, and activation of peripheral immune cells, regulation of cell growth, apoptosis, and modulation of angiostatic effects. It plays an important role in stimulating the activation and migration of immune cells to the site of infection. Binding of CXCL10 to the CXCR3 activates G-protein- mediated signaling and results in downstream activation of phospholipase C-dependent pathways. Activation of the CXCL10/CXCR3 axis also plays an important role in neurons in response to brain injury for activating microglia and directing them to the lesion site. Cleavage of CXCL10 by dipeptidyl peptidase IV (DPP4) removes two amino acids from the N-terminal region that leads to formation of an antagonistic form of CXCL10, a dominant negative form that can bind to CXCR3 but does not induce signaling. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Michlmayr, D., and McKimmie, CS. (2014). Int. J. Interferons, Cytokines, Mediator Res. 6; 1-18; Smit, MJ., et al. (2003). Blood. 102(6); 1959-1965; Angiolillo, AL., et al. (1995). J. Exp. Med. 182(1); 155-162).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service