Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

ZRB1622

Sigma-Aldrich

Anti-RBPMS Antibody, clone 1E16 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Heart and RRM expressed sequence, Hermes, RBP-MS, RNA-binding protein with multiple splicing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E16, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 15.79 kDa (isoform 4
isoform 1)

calculated mol wt 17.02 kDa (isoform 5
isoform 1)

calculated mol wt 21.80 kDa (isoform 1
isoform 1)

calculated mol wt 22.41 kDa (isoform 2
isoform 1)

calculated mol wt 24.28 kDa (isoform 3
isoform 1)

observed mol wt ~18 kDa
observed mol wt ~20 kDa

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... RBPMS(11030)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1E16 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects RNA-binding protein with multiple splicing (RBPMS). It targets an epitope within 21 amino acids from the N-terminal region. May show reactivitry with all isoforms.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the N-terminal region of human RNA-binding protein with multiple splicing (RBPMS).

Application

Anti-RBPMS, clone 1E16 ZooMAb, Cat. No. ZRB1622, is a recombinant Rabbit monoclonal antibody that specifically targets RBPMS and is tested for use in Flow Cytometry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected RBPMS in mouse retina, rat heart, and HepG2 cell lysates.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected RBPMS in one million A549 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected RBPMS in human kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

RNA-binding protein with multiple splicing (UniProt: Q93062; also known as RBP-MS, Heart and RRM expressed sequence, Hermes, RBPMS) is encoded by the RBPMS (also known as HERMES) gene (Gene ID: 11030) in human. RBPMS is a ubiquitously expressed homodimeric protein that acts as a coactivator of transcriptional activity. Its expression level varies depending on the isoform and the tissue type. Its each protein chain binds one RNA molecule via the external surface of the homodimer. It is mainly localized in the nucleus, but it translocates into cytoplasmic stress granules that correspond to P-bodies in response to oxidative stress. It contains a RRM domain (aa 24-101) that is required for its interaction with SMAD4. RBPMS is shown to increase TGF1/SMAD-mediated transactivation. It increases phosphorylation of SMAD2 and SMAD3 on their C-terminal SSXS motif and enhances nuclear accumulation of SMAD proteins. RBPMS also controls alternative RNA splicing of many genes that are essential in smooth muscle cells and any disruption in its regulation results in certain diseases of the heart and blood vessels. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Nakagaki-Silva, EE et al., (2019). eLife. 8; 46327).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service