Skip to Content
MilliporeSigma
All Photos(20)

Key Documents

ZRB1009

Sigma-Aldrich

Anti-CTCF antibody, clone 1B18, ZooMAb® Rabbit monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

11-zinc finger protein, CCCTC-binding factor, CTCFL paralog, Transcriptional repressor CTCF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B18, monoclonal
recombinant monoclonal

description

1B18 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 82.79 kDa
observed mol wt ~120 kDa

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable using 1:100
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable using 1:100
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CTCF(10664)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1B18 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Transcriptional repressor CTCF. It targets an epitope with in 17 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from C-terminal region of human Transcriptional repressor CTCF.

Application

Anti-CTCF, clone 1B18 ZooMAb, Cat. No. ZRB1009, is a recombinant Rabbit Monoclonal Antibody that specifically targets Transcriptional repressor CTCF and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CTCF in NIH3T3 and C6 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CTCF in HUVEC, HeLa, A431, and NIH 3T3 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CTCF in human prostate cancer tissue sections.

Target description

Transcriptional repressor CTCF (UniProt: P49711; also known as 11-zinc finger protein, CCCTC-binding factor, CTCFL paralog) is encoded by the CTCF gene (Gene ID: 10664) in human. CTCF is a ubiquitously distributed chromatin binding factor that binds to DNA sequence specific sites. It contains 11 C2H2-type zinc finger regions. CTCF is involved in transcriptional regulation by binding to chromatin insulators and preventing interaction between promoter and nearby enhancers and silencers. CTCF serves as a transcriptional repressor by binding to promoters of vertebrate MYC gene and BAG1 gene. It can also serve as a transcriptional activator of APP. It also plays a critical role in gene silencing over considerable distances in the genome and preferentially interacts with unmethylated DNA, preventing spreading of CpG methylation and maintaining methylation-free zones. When bound to different DNA sequences, it can dimerize to mediate long-range chromatin looping. CTCF undergoes sumolyation on lysine 74 and 689, which contributes to its repressive function on the MYC P2 promoter. Mutations in CTCF gene are known to cause mental retardation characterized by significantly below average general intellectual functioning associated with impairments in adaptive behavior. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service