Skip to Content
MilliporeSigma
All Photos(12)

Key Documents

ZMS1060

Sigma-Aldrich

Anti-MMP14 Antibody, clone LEM-2/15.8 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

LEM-2/15.8, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 65.89 kDa
observed mol wt ~55 kDa

species reactivity

human

species reactivity (predicted by homology)

rat, monkey, mouse, bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1

epitope sequence

Extracellular domain

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone LEM-2/15.8 is a mouse recombinant monoclonal antibody that specifically detects Matrix metalloproteinase-14 (MMP14). It targets an epitope within 16 amino acids from the extracellular domain.

Immunogen

Linear Peptide corresponding to 16 amino acids from the extracellular domain of human Matrix Metalloproteinase-14 (MMP14).

Application

Quality Control Testing

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected MMP14 in HepG2 cell lysate.

Tested applications

Flow Cytometry Analysis: 1 μg from a representative lot detected MMP14 in one million HepG2 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected MMP14 in human colon tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected MMP14 in HepG2 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-MMP14, clone LEM-2/15.8 ZooMAb®, Cat. No. ZMS1060, is a recombinant Mouse monoclonal antibody that targets MMP14 and is tested for use in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Matrix metalloproteinase-14 (UniProt: P50281; also known as EC:3.4.24.80, MMP-14, MMP-X1, Membrane-type matrix metalloproteinase 1, MT-MMP 1, MTMMP1, Membrane-type-1 matrix metalloproteinase, MT1-MMP, MT1MMP) is encoded by the MMP14 gene (Gene ID: 4323) in human. MMPs are a family zinc-dependent endoproteinases that degrade collagen and other structural proteins and are responsible for much of the turnover of matrix components. Each MMP contains a protease domain with a conserved HExGHxxGxxHS/T sequence, where three histidine residues form a complex with a catalytic Zinc atom. A regulatory domain with a conserved PRCGxPD motif helps to maintain MMPs in their latent form by binding of the cysteine residue to the active site Zn. MMP-14 is a single-pass type I membrane protein that is expressed in stromal cells of colon, breast, head, and neck. It is a membrane type matrix metalloproteinase (MT-MMP) that is synthesized with a signal peptide (aa 1-20) and a propeptide (aa 21-111) that are subsequently cleaved off to generate the active mature form. The propeptide maintains it in a latent state. The mature enzyme contains an extracellular domain (aa 112-541), a transmembrane domain (aa 542-562), and a cytoplasmic domain (aa 563-582). MMP14 contains four hemopexin domains within its extracellular domain and a cysteine switch (aa 91-98). The conserved cysteine present in the cysteine-switch motif binds the catalytic zinc ion, thus inhibiting the enzyme. The dissociation of the cysteine from the zinc ion upon the release of propeptide activates the enzyme. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service