Skip to Content
MilliporeSigma
All Photos(10)

Key Documents

ZMS1046

Sigma-Aldrich

Anti-Rabies glycoprotein Antibody, clone 24-3F-10 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

24-3F-10, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 58.66 kDa

purified by

using protein G

species reactivity

virus

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable

isotype

IgG2a

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 24-3F-10 is a ZooMAb® Mouse recombinant monoclonal antibody that detects glycoprotein of Rabies virus.

Immunogen

Protein G of Evelyn Rokitniki Abelseth(ERA) strain of Rabies virus.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in L929 cells infected with Rabies virus.

Immunocytochemistry Analysis (ICC): A 1:100 dilution of this antibody detected Rabies virus glycoprotein in L929 cells infected with Rabies virus.

Tested applications

Immunofluorescence Analysis: A representative lot detected Rabies glycoprotein in Immunofluorescence applications (Do-Phong, Johnny., et. al. (2016). Elife. 5:e13214).

Flow Cytometry Analysis: 0.1 μg from a representative lot detected Rabies glycoprotein in one million L929 cells infected with Rabies virus.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Rabies glycoprotein, clone 24-3F-10 ZooMAb®, Cat. No. ZMS1046, is a recombinant Mouse monoclonal antibody that detects Rabies virus glycoprotein and is tested for use in Flow Cytometry, Immunocytochemistry, and Immunofluorescence.

Target description

Glycoprotein (UniProt: P03524) is encoded by the G gene in Rabies virus, strain ERA. Glycoprotein is a single-pass type I membrane glycoprotein that belongs to the lyssavirus glycoprotein family. It attaches the virus to host cellular receptor, inducing endocytosis of the virion. In the endosome, the acidic pH induces conformational changes in the glycoprotein trimer, which trigger fusion between virus and cell membrane. The muscle form of the nicotinic acetylcholine receptor (nAChR), the neuronal cell adhesion molecule (NCAM), and the p75 neurotrophin receptor (p75NTR) are reported to bind glycoprotein and thereby facilitate rabies virus entry into cells. Glycoprotein is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off in the mature form. Its structure includes a virion surface (aa 20-459), helical region (aa 460-480) and an intravirion (aa 481-524). Arginine 352 is reported to be involved in Rabies virus pathogenicity and its mutation attenuates the virus. It is glycosylated and palmitoylated by host and its glycosylation is considered to be crucial for its export at the cell surface. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service