Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ZFNSLC35A1

SAFC

CompoZr® Knockout ZFN Kit- CHO SLC35A1

Synonym(s):

CompoZr® Knockout ZFN Kit for CHO Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

description

Solute Carrier Family 35 (CMP-Sialic Acid Transporter), Member A1

Quality Level

product line

CompoZr®

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

CompoZr Knockout ZFNs create targeted double strand breaks at a gene specific target locus. Through the cellular process of Non-Homologous End Joining, this double strand break can result in modification of the DNA sequence and therefore create a functional knockout of the targeted gene. To provide the best chance of creating a functional gene knockout, CompoZr Knockout ZFNs are targeted to the first 2/3 of the coding region for the gene of interest.

Physical form

CompoZr Knockout ZFNs are provided as 10 aliquots of mRNA encoding for the ZFNs along with plasmid DNA for both ZFN pairs. Also included in the kit are custom-designed primers, that allow for determination of the rate of mutation and for screening of individual clones that have the desired mutational event, and positive control DNA that has been modified using the CompoZr Knockout ZFN.

Legal Information

All Zinc Finger Nucleases are sold under license from Sangamo Biosciences. For a copy of the Label License provided with purchase of CompoZr ZFNs, please visit the ZFN Label License page.
CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn more about cell line development and the relevant products you need for this process.

Learn more about cell line development and the relevant products you need for this process.

Learn more about cell line development and the relevant products you need for this process.

Learn more about cell line development and the relevant products you need for this process.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service