Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z744120

Freezer box

W × L 2 in. × 2 in., to hold, 100 tube(s)

Synonym(s):

freezer container, freezer storage, storage container

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123300

material

white

feature

to hold 100 tube(s)

packaging

pack of 1 ea

manufacturer/tradename

Diversified Biotech FRSB-2000

technique(s)

cryopreservation: suitable

W × L

2 in. × 2 in.

Looking for similar products? Visit Product Comparison Guide

General description

2" Freezer Box w/100 hole divider

Application

Cryogenics

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service