Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z744109

CoolSafe box

Synonym(s):

Cooling Box, dry block, tube cooling block, cooling block

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123300

packaging

pack of 1 ea

manufacturer/tradename

Diversified Biotech CSF-BOX

I.D. × L × H

6.25 in. × 3.25 in. × 4.62 in.

O.D. × L × H

8.5 in. × 5.5 in. × 6.5 in.

General description

The CoolSafe box is made of triple density expanded polystyrene and molded under extreme pressure creating a durable container that guarantees temperature stability.

Application

PCR

Legal Information

CoolSafe is a trademark of Diversified Biotech, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service