Key Documents
Z742657
Tube-O-Reactor
Medi, MWCO 8 kDa
About This Item
UNSPSC Code:
41123100
NACRES:
NB.35
feature
Medi
packaging
pkg of 5 ea
manufacturer/tradename
G-BioSciences 786-027-8K
pore size
8 kDa MWCO
General description
Tube-O-Reactor is a system that allows all the key steps of cross linking, coupling and modification of proteins and/or nucleic acids to be performed in a single tube. This minimizes the risk of sample loss, of experimental time and of hands-on phases.
The Tube-O-Reactor system is available in three MWCO sizes, 4kDa, 8kDa and 15kDa. Tube-O-Reactoris supplied as a Micro kit for sample sizes of 20-250μl and a Medi size for samples of 0.2-2ml.
Each Tube-O-Reactoris suitable for 5 reactions, depending on sample volumes, and is supplied with:
  • 5 Medi or 5 Micro Tube-O-Reactor
  • 5 Floats for each size of Tube-O-Reactor
  • 5 Micro Dialysis Reaction Chambers
  • Glass balls
Legal Information
Tube-O-Reactor is a trademark of Geno Technology, Inc.