Key Documents
Z740505
Levgo® smartSpatula® disposable PP laboratory spatula, Look for the V at the neck
L 140 mm, white, Biodegradable EcoSpatula®, short
Synonym(s):
biodegradable spatula, disposable spatula, microspatula
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
material
polypropylene
white
packaging
pack of 300 ea
case of 7200 ea
manufacturer/tradename
LevGo Inc 17261
L
140 mm
General description
Versatile Double-ended polypropylene tool that can scoop, scrape, spoon, stir, collect, transfer, handle or manipulate a variety of chemicals and biological materials.
  • The V design at the neck of the disposable lab spatula is atrademark of LevGo, Inc.
  • No Searching, washing or contamination.
  • ~99% Polypropylene and ~ 1% EcoPure additive*, is autoclavable and resistant to dilute acids and bases.
  • Polypropylene will not shatter after exposure to liquid nitrogen frozen samples.
  • Hollow core uses less plastic than other spatulas and minimizes heat or cold transfer.
  • SGS-Tested for US and EU food contact standards.
  • Sustainability – LevGo plants a tree for every case sold.
LevGo spatulas are widely used in commercial, university, diagnostic and pharmaceutical labs worldwide as well as by compounding pharmacists to produce customized medications.
ecoSpatulas are 99% polypropylene (PP) resin and 1% EcoPure organic additive, which helps promote biodegradation of PP. Representative samples of our ecoSpatulas and our standard 100% PP spatulas were tested according to the ASTM D5511 test method (intended to replicate conditions in biologically active landfills). ecoSpatulas biodegraded an average of 29% in the first 586 days of testing compared to 8% for standard 100% PP spatulas. Tests were conducted using 48% solids content, which is not typical; solids content in naturally wetter landfills range from 55% to 65%, while the driest landfills may reach 93%. Actual biodegradation rates will vary in biologically active landfills according to the solids content, temperature, and moisture level of the landfill. ecoSpatulas retain all the original properties and shelf life of our standard PP spatulas: they are autoclavable, resistant to dilute acids and bases, and FDA compliant for food contact. They will not degrade during use. It is only after extended exposure to the microbes in a biologically active landfill that they begin to break down and eventually turn into inert humus and natural gases. Important Notice: Laws in California, Maryland, and Washington prohibit the sale of certain products and packaging that are labeled with the terms “biodegradable”, “degradable”, or “decomposable” or any form of those terms, or that imply in any way that the item will break down, biodegrade, or decompose in a landfill or other environment. These restrictions apply to all sales in or into these states, including such sales over the Internet. Our ecoSpatula product packaging meets the requirements of California, Maryland, and Washington.
Legal Information
EcoPure is a registered trademark of Bio-Tec Environmental, LLC
LevGo is a registered trademark of LevGo Inc.
Look for the V at the neck is a trademark of LevGo Inc.
ecoSpatula is a registered trademark of LevGo Inc.
smartSpatula is a registered trademark of LevGo, Inc.