Key Documents
Z721646
Zone-Free Sealing Films
non-sterile
Synonym(s):
qPCR plate seal
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
material
acrylic adhesive
polyethylene film (top layer)
white polypropylene film (sublayer)
sterility
non-sterile
feature
piercable
packaging
pack of 50 ea
manufacturer/tradename
Excel Scientific ZAF-PE-50
L × W
79.4 mm × 142.9 mm
General description
With a 70 μm polyethylene top layer and inert white polypropylene and acrylic adhesive sublayer, this easy-piercing sealing film allows direct sample recovery with single or multichannel pipettors and robotic probes. The adhesive-free zone above each well prevents adhesive fouling of the pipet tip or probe, ultimately minimizing the amount of adhesive that comes into contact with the sample. Dimensions are 79.4 by 142.9 mm for 96-well plates.
Features and Benefits
  • clear zone above each well is free of adhesive and easily pierceable
  • clear zones and end tabs facilitate well alignment and accurate positioning
  • inert, chemical resistant
  • functional temperature range -40 C to +90 C
Legal Information
Zone-Free is a trademark of Excel Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.
Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.
Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.
Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.
See All