Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z709328

ART® self-sealing barrier pipette tips

volume range 100-1000 μL, sterile, ART 1000

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier

packaging

pack of 800 ea

manufacturer/tradename

Molecular BioProducts 2279

technique(s)

PCR: suitable

maximum volume

1 mL

volume range

100-1000 μL

Looking for similar products? Visit Product Comparison Guide

General description

Achieve precision in your applications with ART® self-sealing barrier pipette tips. These tips effectively seal when encountering potential contaminants, securely trapping them within the barrier. Designed to support various pipetting needs, ART barrier specialty tips are suitable for a wide range of applications.

Application

ART® self-sealing barrier pipette tips are suitable for use in:
  • Forensics
  • General PCR work
  • Radioisotope sampling
  • Genetic studies

ART® self-sealing barrier pipette tips have been used for the quantitative profiling of endocannabinoids in lipoproteins by LC–MS (liquid chromatography–mass spectrometry)/MS. They have also been used for RNA isolation and reverse transcription.

Features and Benefits

  • Free of DNase, RNase, DNA, ATP, PCR inhibitors, and endotoxins.
  • Available in a wide range of options including sterile, non-sterile, and various packaging options.
  • Manufactured in compliance with the industry′s highest quality standards.

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Compatibility

Compatibility
Biohit mLINE 100-1000 μl
Biohit Proline-Electronic 1000 μl
Biohit Variable 200-1000 μl
Capp Aero 1000 μl
Corning 4964 100-1000 μl
Eppendorf Research/Reference 100-1000 μl
Eppendorf 4810 100-1000 μl
Eppendorf Research Pro 50-1000 μl
Eppendorf Response 4850 50-1000 μl
Finnpipette 4027/4500 Digital 200-1000 μl
Mettler-Toledo VoluMate 100-1000 μl
Pipetman P-1000 100-1000 μl
Rainin EDP 2 100-1000 μl
Rainin EDP Plus 100-1000 μl
Rainin Pipet Plus 100-1000 μl
Rainin Pipetman 100-1000 μl
Transferpette 100-1000 μl

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro culture and analysis of adult hippocampal neural progenitors.
Peltier J, et al.
Methods in Molecular Biology, 621, 65-87 (2010)
Quantitative profiling of endocannabinoids in lipoproteins by LC-MS/MS.
Bilgin M, et al.
Analytical and Bioanalytical Chemistry, 407, 5125-5131 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service