Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z709131

ART® self-sealing barrier pipette tips

ART Gel 20P, volume range 0.5-20 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier

packaging

pack of 960 ea (10 racks of 96 tips)

manufacturer/tradename

Molecular BioProducts 2155P

technique(s)

PCR: suitable

maximum volume

20 μL

volume range

0.5-20 μL

Looking for similar products? Visit Product Comparison Guide

General description

Perfect for low-volume gel loading

Application

ART® self-sealing barrier pipette tips have been used in PCR reactions and LC–MS/MS (liquid chromatography–mass spectrometry).

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Compatibility

Compatibility
Biohit Proline-Electronic 10-100 μl
Eppendorf 4810 2-20 μl
Eppendorf Research/Reference 2-20 μl
Finnpipette 4500 Digital .5-10 μl
Hamilton SoftGrip Precision 25 μl
Mettler-Toledo VoluMate 2-20 μl
Nichriyo/Oxford BenchMate 2-20 μl
Pipetman P-20 2-20 μl
Rainin EDP 2 2.5-25 μl
Rainin EDP Plus 2.5-25 μl
Rainin Pipet Plus 2.5-20 μl
Rainin Pipetman 2.5-20 μl
Socorex/Wheaton Calibra/822 2-20 μl
Transferpette 2-20 μl
Transferpette Multi-Channel 5-50 μl

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prolonged arthritis associated with sindbis-related (Pogosta) virus infection.
Laine M
Rheumatology (Oxford, England), 39, 1272-1274 (2000)
Nitrogenase genes in PCR and RT-PCR reagents: implications for studies of diversity of functional genes.
Zehr JP
Biotechniques, 35, 996-1002 (2003)
Quantitative profiling of endocannabinoids in lipoproteins by LC-MS/MS.
Bilgin M, et al.
Analytical and Bioanalytical Chemistry, 407, 5125-5131 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service