Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z688487

Bio-Pure alcohol wipes

100% biodegradable

Synonym(s):

cleaning wipes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131905
NACRES:
NB.35

manufacturer/tradename

Diversified Biotech BPWP-1000

packaging

40 ea of

General description

Proprietary fabric used to manufacture Bio-Pure alcohol wipes is formulated to easily break down over time and is 100% biodegradable. The re-sealable and flexible packaging has a low carbon footprint.

Features and Benefits

  • 100% Biodegradable with re-sealable packaging reduces waste, maintaining product longevity and reducing environmental impact.
  • Formulated with 70% isopropanol alcohol and 30% water to guarantee effective cleaning and disinfection.
  • Strong, lint-free wipes protect delicate surfaces, ensuring precision cleaning without leaving behind particles or scratches
  • Durable, strong fabric construction prevents the wipes from tearing or falling apart during use, enhancing convenience and reliability in routine lab cleaning tasks.

Legal Information

Bio-Pure is a trademark of Diversified Biotech

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service