Key Documents
Z328820
BRAND® magnetic stirring bar, PTFE, cylindrical
L 45 mm, bar diam. 8 mm
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
material
PTFE-coated
cylindrical
manufacturer/tradename
BRAND 137132
L
45 mm
bar diam.
8 mm
General description
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
Features and Benefits
  • BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
  • The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
  • Ideal for large volumes or highly viscous mixtures.
Other Notes
BR137132 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG