Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4039

Sigma-Aldrich

Fgf-2 from rat

recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)

Synonym(s):

FGB-b, FGF-2, Fibroblast Growth Factor-basic, HBGF-2, Prostatropin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

Assay

≥97% (HPLC)
≥97% (SDS-PAGE)

form

lyophilized

potency

<0.2 ng/mL

mol wt

~16.3 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Fgf2(54250)

General description

Fibroblast growth factor-2 (FGF-2) is a basic heparin binding growth factor and a profibrotic factor. The recombinant mouse FGF-2 is a single, non-glycosylated chain containing 145 amino acids and having a molecular weight of 16.3kDa.

Biochem/physiol Actions

Fibroblast growth factor-2 (FGF-2) stimulates the proliferation of a wide variety of cells including mesenchymal, neuroectodermal and endothelial cells.

Physical form

Lyophilized from 1 mg/ml solution after extensive dialysis against 20 mM phosphate buffer, pH 7.4 and 130 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in sterile water to 0.1–1 mg/mL.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

FGF-18, a novel member of the fibroblast growth factor family, stimulates hepatic and intestinal proliferation.
Hu MC
Molecular and Cellular Biology, 18(10), 6063-6074 (1998)
Apocynin Attenuates Cardiac Injury in Type 4 Cardiorenal Syndrome via Suppressing Cardiac Fibroblast Growth Factor-2 With Oxidative Stress Inhibition.
Liu Y
Journal of the American Heart Association, 4(7), e001598-e001598 (2015)
Larisa Antonova et al.
Pharmaceuticals (Basel, Switzerland), 14(4) (2021-04-04)
Tissue-engineered vascular grafts are widely tested as a promising substitute for both arterial bypass and replacement surgery. We previously demonstrated that incorporation of VEGF into electrospun tubular scaffolds from poly(3-hydroxybutyrate-co-3-hydroxyvalerate)/poly(ε-caprolactone) enhances formation of an endothelial cell monolayer. However, an overdose
Yayoi Yoshimura et al.
PloS one, 10(6), e0128288-e0128288 (2015-06-04)
Glioblastoma multiforme (GBM) is associated with high mortality due to infiltrative growth and recurrence. Median survival of the patients is less than 15 months, increasing requirements for new therapies. We found that both arsenic trioxide and 10058F4, an inhibitor of
Shun Yamamuro et al.
International journal of oncology, 47(1), 91-96 (2015-05-09)
Glioma stem-like cells (GSCs) could have potential for tumorigenesis, treatment resistance, and tumor recurrence (GSC hypothesis). However, the mechanisms underlying such potential has remained elusive and few ultrastructural features of the cells have been reported in detail. We therefore undertook

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service