Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0541

Sigma-Aldrich

SIRP-alpha (CD172A), His-Tag

recombinant, expressed in HEK 293 cells

Synonym(s):

Bit, CD172a, MyD-1, Tyrosine-protein phosphatase non-receptor type substrate 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

38.4 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SIRPA(140885)

General description

Signal regulatory protein-alpha (SIRP-α), also known as Tyrosine-protein phosphatase non-receptor type substrate 1, CD172a, Bit, and MyD-1, GenBank Accession No. NP_542970, a.a. 27-370, with C-terminal His-tag, MW 38.4 kDa*, expressed in a HEK293 expression system. This protein acts as a receptor for CD47; a.a. 27-370 correspond to the extracellular portion of SIRP-α. *Note: SIRP-α is heavily glycosylated, resulting in apparent higher molecular weight.

Physical form

Aqueous solution

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service