Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0440

Sigma-Aldrich

UBE1 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE)

Synonym(s):

UBA1, UBA1A, ubiquitin-like modifier activating enzyme 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

118 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... UBA1(7317)

Related Categories

General description

Human UBE1 (GenBank Accession No. NM_003334) amino-acids 2-end with N-terminal FLAG-tag, MW=118 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful in conjunction with E2 and E3 for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Hui Li et al.
Experimental cell research, 371(1), 250-254 (2018-08-18)
Ninety percent of all cancer related deaths happen due to metastatic progression. One important protein facilitating metastatic progression in hepatocellular carcinoma (HCC) is the metastasis associated 1 protein (MTA-1). We have earlier shown that in the context of HCC and
Yi Liu et al.
Journal of experimental botany, 71(18), 5562-5576 (2020-05-27)
Plants can be simultaneously exposed to multiple stresses. The interplay of abiotic and biotic stresses may result in synergistic or antagonistic effects on plant development and health. Temporary drought stress can stimulate plant immunity; however, the molecular mechanism of drought-induced
UBE1 expression in extranodal NK/T cell lymphoma, nasal type.
Z L Su et al.
Leukemia & lymphoma, 49(9), 1821-1822 (2008-07-29)
Juliane Ramser et al.
American journal of human genetics, 82(1), 188-193 (2008-01-09)
X-linked infantile spinal muscular atrophy (XL-SMA) is an X-linked disorder presenting with the clinical features hypotonia, areflexia, and multiple congenital contractures (arthrogryposis) associated with loss of anterior horn cells and infantile death. To identify the XL-SMA disease gene, we performed
J Romaní-Aumedes et al.
Cell death & disease, 5, e1364-e1364 (2014-08-08)
Mutations in the PARK2 gene are associated with an autosomal recessive form of juvenile parkinsonism (AR-JP). These mutations affect parkin solubility and impair its E3 ligase activity, leading to a toxic accumulation of proteins within susceptible neurons that results in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service