Skip to Content
MilliporeSigma
All Photos(2)

Documents

SRE0010

Sigma-Aldrich

5-Methyltetrahydrofolic acid disodium salt

≥88% (UV-vis), Suitable for manufacturing of diagnostic kits and reagents

Synonym(s):

5-Methyltetrahydrofolic acid disodium salt, 5-Methyl-5,6,7,8-tetrahydropteroyl-L-glutamic acid disodium salt, 5-Methyl-THF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23N7Na2O6
CAS Number:
Molecular Weight:
503.42
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥88% (UV-vis)

form

powder

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].CN1C(CNc2ccc(cc2)C(=O)N[C@H](CCC([O-])=O)C([O-])=O)CNc3nc(N)nc(O)c13

InChI

1S/C20H25N7O6.2Na/c1-27-12(9-23-16-15(27)18(31)26-20(21)25-16)8-22-11-4-2-10(3-5-11)17(30)24-13(19(32)33)6-7-14(28)29;;/h2-5,12-13,22H,6-9H2,1H3,(H,24,30)(H,28,29)(H,32,33)(H4,21,23,25,26,31);;/q;2*+1/p-2

InChI key

KKIWVYLOTHCGRV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

5-Methyl-THF acts as both cofactor and substrate in the regeneration of methionine from homocysteine through the action of 5-methyl-THF-homocysteine S-methyltransferase.

Packaging

Sealed ampule.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service