Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3984

Sigma-Aldrich

Insulin sensitizer, C59

new

≥98% (HPLC)

Synonym(s):

N-(4,5-Dichloro-2-pyridinyl)tricyclo[3.3.1.13,7]decane-1-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18Cl2N2O
CAS Number:
Molecular Weight:
325.23
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Orally available potentiator of insulin action that significantly improves glucose tolerance in DIO mice; restores insulin-stimulated GLUT4 translocation.



Insulin sensitizer C59 is an orally available potentiator of insulin action that significantly improves glucose tolerance in DIO mice. Insulin sensitizer C59 restores insulin-stimulated GLUT4 translocation without targeting PPARγ. It acts through interaction with Unc119 proteins. C59 lowers fasting blood glucose and plasma insulin concentrations in in DIO mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Terry C Yin et al.
Cell chemical biology, 30(8), 933-942 (2023-07-16)
Insulin resistance (IR) is the root cause of type II diabetes, yet no safe treatment is available to address it. Using a high throughput compatible assay that measures real-time translocation of the glucose transporter glucose transporter 4 (GLUT4), we identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service