Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3214

Sigma-Aldrich

LDC7559

≥98% (HPLC)

Synonym(s):

LDC 7559, N-[2-(2-Methoxyphenyl)-4H,10H-pyrazolo[5,1-c][1,4]benzoxazepin-7-yl]-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19N3O3
CAS Number:
Molecular Weight:
349.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

LDC7559 is a potent and selective activator of PFKL (phosphofructokinase-1 liver type) that suppress excessive NOX2-dependent oxidative burst and neutrophil cell death (NETosis). LDC7559 decreases neutrophiles efficiency to kill bacteria. It prevents neutrophil-induced tissue damage.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service