Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2886

Sigma-Aldrich

O4I3

≥98% (HPLC)

Synonym(s):

2-(4-Methylphenyl)-6-methylimidazo[1,2-a]pyridine, 6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine, OCT4-inducing compound 3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N2
CAS Number:
Molecular Weight:
222.29
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=CC=C(C2=CN3C=C(C=CC3=N2)C)C=C1

InChI

1S/C15H14N2/c1-11-3-6-13(7-4-11)14-10-17-9-12(2)5-8-15(17)16-14/h3-10H,1-2H3

InChI key

AWEWSJJCANQFRB-UHFFFAOYSA-N

Biochem/physiol Actions

O4I3 is a highly potent and KDM5A (JARID1A) subtype-selective H3K4 demethylase KDM5 inhibitor (KDM5A IC50 = 0.15 nM vs KDM5B/C/D IC50 = 125.2/9.76/4.7 nM, KDM4 IC50 = 249 nM, LSD1 IC50 >100 μM) that effectively induces OCT4 expresson and improves OSKM-induced pluripotent stem cell (iPSC) reprogramming efficiency of human primary fibroblasts (50 nM) by preventing KDM5A from interfering with H3K4Me3 enrichment at the OCT4 promoter. O4I3 promotes hPSC homeostasis and represses cellular differentiation, increasing hPSCs viability up to 4-fold at a low 10 nM concentration when compared with Rock inhibitor Y-27632 treatment at 10 μM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasamin Dabiri et al.
iScience, 12, 168-181 (2019-01-28)
Pioneering human induced pluripotent stem cell (iPSC)-based pre-clinical studies have raised safety concerns and pinpointed the need for safer and more efficient approaches to generate and maintain patient-specific iPSCs. One approach is searching for compounds that influence pluripotent stem cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service