Skip to Content
MilliporeSigma

SML1306

S-p-Bromobenzylglutathione cyclopentyl diester

≥98% (HPLC), powder, Glyoxalase 1 inhibitor

Synonym(s):

BBGC, BBGD, BrBzGCp2, N-[S-[(4-Bromophenyl)methyl]-N-L-γ-glutamyl-L-cysteinyl]-Glycine dicyclopentyl ester, S-p-Bromobenzylglutathione dicyclopentyl ester, S-para-Bromobenzylglutathione cyclopentyl diester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H38BrN3O6S
CAS Number:
Molecular Weight:
612.58
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

S-p-Bromobenzylglutathione cyclopentyl diester, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

BrC1=CC=C(CSC[C@H](NC(CC[C@H](N)C(OC2CCCC2)=O)=O)C(NCC(OC3CCCC3)=O)=O)C=C1

InChI

1S/C27H38BrN3O6S/c28-19-11-9-18(10-12-19)16-38-17-23(26(34)30-15-25(33)36-20-5-1-2-6-20)31-24(32)14-13-22(29)27(35)37-21-7-3-4-8-21/h9-12,20-23H,1-8,13-17,29H2,(H,30,34)(H,31,32)/t22-,23-/m0/s1

InChI key

QIFSPGPRHFNZNN-GOTSBHOMSA-N

Application

S-p-Bromobenzylglutathione cyclopentyl diester has been used as an inhibitor of glyoxalase 1 (GLO-1) enzyme to treat HL-1 cardiomyocytes for stimulating aging-based glycative stress. It has also been used as an inhibitor of GLO-1 in hepatic stellate cells.

Biochem/physiol Actions

S-p-Bromobenzylglutathione cyclopentyl diester (BBGC) is a cell-permeable inhibitor of Glyoxalase 1 (GLO1), an enzyme that detoxifies methylglyoxal, a toxic side-product of glycolysis that induces apoptosis at high levels and has been linked to arterial atherogenesis in diabetics. However, tumors use GLO as well and overexpression of GLO1 has been reported in a number of cancers, so GLO1 inhibitors including BBCG have been investigated as possible anticancer agents. Recent studies have indicated that methylglyoxal has some beneficial effects at low levels with GABAA receptor agonist activity, and that GLO1 inhibition with BBGC can reduce anxiety-like behavior and decrease antiseizure activity. S-p-bromobenzylglutathione cyclopentyl diester should be an inportant tool to study the glyoxalase system.
S-p-Bromobenzylglutathione cyclopentyl diester (BBGC) is a cell-permeable inhibitor of Glyoxalase 1 (GLO1).
BBGC possesses antiproliferative activity, observed in human leukemia cells.
Glyoxalases are associated with detoxification process. Inhibition of glyoxalases produces an anti-carcinogenic and anti-inflammatory effect.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library