Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1205

Sigma-Aldrich

BVT.2733

≥98% (HPLC)

Synonym(s):

1-[[2-[[(3-Chloro-2-methylphenyl)sulfonyl]amino]-4-thiazolyl]acetyl]-4-methyl-piperazine, 3-Chloro-2-methyl-N-[4-[2-(4-methyl-1-piperazinyl)-2-oxoethyl]-2-thiazolyl]-benzenesulfonamide, BVT 2733

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21ClN4O3S2
CAS Number:
Molecular Weight:
428.96
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(C1=C(C)C(Cl)=CC=C1)(NC2=NC(CC(N3CCN(C)CC3)=O)=CS2)=O

InChI

1S/C17H21ClN4O3S2/c1-12-14(18)4-3-5-15(12)27(24,25)20-17-19-13(11-26-17)10-16(23)22-8-6-21(2)7-9-22/h3-5,11H,6-10H2,1-2H3,(H,19,20)

InChI key

YDPRNGAPPNPYQQ-UHFFFAOYSA-N

Biochem/physiol Actions

BVT.2733 is a potent and selective inhibitor of 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) that protects osteoblasts against endogenous glucocorticoid induced dysfunction. BVT.2733 lowers blood glucose levels in in hyperglycaemic mice.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service