Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0943

Sigma-Aldrich

YM-1

Synonym(s):

2-[[3-Ethyl-5-(3-methyl-2(3H)-benzothiazolylidene)-4-oxo-2-thiazolidinylidene]methyl]-1-methyl-pyridinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20ClN3OS2
CAS Number:
Molecular Weight:
417.98
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

color

orange to dark red

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Application

YM-1 has been used as a Hsp70 (heat shock protein) inhibitor in cell viability assay.

Biochem/physiol Actions

YM-1 modulates Hsp70 activity by binding to the nucleotide-binding domain of ADP- but not ATP-bound Hsp70, stabilizing Hsp70 in its ADP-bound conformation. The ADP-bound state has tight affinity for its substrates, promoting their ubiquitination and degradation, and a greatly reduced off rate compared to the ATP-bound state. YM-1 appears to behave in a similar manner to Hip, a Hsp70 co-chaperone that stabilizes Hsp70 in its ADP-bound conformation which favors degradation of misfolded substrates such as those involved in various neurodegenerative diseases.

YM-1 also has anticancer activity, which could be due to enhanced ubiquitination and clearance of Akt, a major survival kinase. In studies with multiple cancer lines, YM-1 was found to selectively target cancer cells over normal cells and to overcome tamoxifen resistance in a tamoxifen-resistant MCF7 cell model. YM-1 had greater efficacy and cytosolic localization than the closely related MKT-077 (M5449).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Effect of an Inhibitor of HSP70, YM-1, on Hikeshi Knockout Cells.
RAHMAN K Z, et al.
Japanese Society for Thermal Medicine, 33(4), 117-122 (2017)
Chieh-Ting Fang et al.
Cell death & disease, 11(8), 715-715 (2020-09-03)
The heat shock protein 70 (HSP70) is a conserved molecular chaperone and proteostasis regulator that protects cells from pharmacological stress and promotes drug resistance in cancer cells. In this study, we found that HSP70 may promote resistance to anticancer drugs
Ayushman Ghosh et al.
Intervirology, 60(6), 247-262 (2018-06-29)
The study was designed to identify putative Chikungunya virus (CHIKV) receptor/s on C6/36 cells that facilitate viral entry. The virus overlay protein binding assay (VOPBA) was adopted to identify CHIKV-interacting bands present in C6/36 cell membrane and identity of the

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service