Skip to Content
MilliporeSigma

SML0585

Piclamilast

≥98% (HPLC)

Synonym(s):

3-(Cyclopentyloxy)-N-(3,5-dichloro-4-pyridinyl)-4-methoxy-benzamide, N-(3,5-Dichloropyrid-4-yl)-3-cyclopentyloxy-4-methoxybenzamide, RP 73-401, RP 73401, RPR 73401

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H18Cl2N2O3
CAS Number:
Molecular Weight:
381.25
UNSPSC Code:
41106305
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1cncc(c1NC(=O)c2cc(c(cc2)OC)OC3CCCC3)Cl

InChI

1S/C18H18Cl2N2O3/c1-24-15-7-6-11(8-16(15)25-12-4-2-3-5-12)18(23)22-17-13(19)9-21-10-14(17)20/h6-10,12H,2-5H2,1H3,(H,21,22,23)

InChI key

RRRUXBQSQLKHEL-UHFFFAOYSA-N

Application

Piclamilasthas been used as a phosphodiesterase-4 (PDE4) inhibitor to treat cultures at the indicated time points after blast exposure.

Biochem/physiol Actions

Piclamilast is a potent and selective cyclic AMP phosphodiesterase-4 inhibitor that exhibits equal, high affinity for both the PDE4 high- and low-affinity rolipram binding states (HARBS and LARBS). Recent studies shown that piclamilast inhibits Trypanosoma brucei PDEB1 (TbrPDEB1) and TbrPDEB2.
Piclamilast is a potent and selective cyclic AMP phosphodiesterase-4 inhibitor.
Piclamilast is also known as N-(3,5-dichloropyrid-4-yl)-3-cyclopentyloxy-4-methoxybenzamide. Piclamilast regulates the cAMP (cyclic adenosine monophosphate) signaling pathway. It increases the retinoid-dependent transactivation and the degradation of retinoic acid receptor α (RARα).

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphodiesterases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library