Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML0507

Sigma-Aldrich

NPC26

≥98% (HPLC)

Synonym(s):

4,5,6,7-Tetrahydro-5,5,7,7-tetramethyl-2-[[(5-nitro-2-thienyl)carbonyl]amino]-thieno[2,3-c]pyridine-3-carboxylic acid ethyl ester, NPC-26

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23N3O5S2
CAS Number:
Molecular Weight:
437.53
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

light orange to dark orange

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CCOC(=O)c1c2CC(C)(C)NC(C)(C)c2sc1NC(=O)c3ccc(s3)[N+]([O-])=O

InChI

1S/C19H23N3O5S2/c1-6-27-17(24)13-10-9-18(2,3)21-19(4,5)14(10)29-16(13)20-15(23)11-7-8-12(28-11)22(25)26/h7-8,21H,6,9H2,1-5H3,(H,20,23)

InChI key

DWGSXNATVAGHKZ-UHFFFAOYSA-N

Biochem/physiol Actions

NPC26 induces mitochondrial fragmentation and cell death in RAS expressing cells, similar to erastin. The effects of NPC26 are independent of ROS scavengers, which can block cell death induced by erastin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service