Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB00208

Sigma-Aldrich

N-trans-caffeoyltyramine

≥85% (LC/MS-ELSD)

Synonym(s):

Typheramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17NO4
CAS Number:
Molecular Weight:
299.32
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥85% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

Oc1ccc(CCNC(=O)\C=C\c2ccc(O)c(O)c2)cc1

InChI

1S/C17H17NO4/c19-14-5-1-12(2-6-14)9-10-18-17(22)8-4-13-3-7-15(20)16(21)11-13/h1-8,11,19-21H,9-10H2,(H,18,22)/b8-4+

InChI key

VSHUQLRHTJOKTA-XBXARRHUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William Leonard et al.
Food chemistry, 346, 128606-128606 (2021-01-04)
The impact of extrusion at different barrel temperature and screw speed on the hempseed hull was investigated. The extrusion treatments showed significant (p < 0.05) increase in total phenolic content, proportion of free phenolic compounds, and DPPH and ABTS radical scavenging activities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service