Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0252

Sigma-Aldrich

Thrombin Substrate, fluorogenic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H42N7O7
Molecular Weight:
624.71
UNSPSC Code:
12352202
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

BOC-Val-Pro-Arg-AFC

General description

Thrombin has procoagulant and anticoagulant properties. It also has chemotactic and mitogenic effects. Thrombin triggers the growth of mammalian cells, fibroblasts and macrophage- like tumor cell lines. It regulates brain development. Thrombin plays an important role in hemostasis and thrombosis. It is associated with mast cell degranulation along with the release of histamine and enhances the production of IL-1 (interleukin-1) by activated macrophages. Thrombin stimulates vascular permeability and transendothelial cell migration by neutrophils.

Application

Thrombin is a serine protease involved in the coagulation cascade wherein it converts fibrinogen into fibrin. Thrombin may be detected (assayed) using peptides that contain the VPR sequence such as the fluorogenic substrates Boc-VPR-AFC, Boc-Val-Pro-Arg-7-AMC and the chromogenic substrate Boc-Val-Pro-Arg-pNA. These substrates may also be useful to detect other serine proteases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thrombin functions as an inflammatory mediator through activation of its receptor.
Cirino G, et al.
The Journal of Experimental Medicine, 183(3), 821-827 (1996)
Multifunctional roles of thrombin
Narayanan S
Annals of Clinical and Laboratory Science, 29(4), 275-280 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service