Skip to Content
MilliporeSigma
All Photos(1)

Documents

SBR00012

Sigma-Aldrich

Lipid A, monophosphoryl Salmonella enterica serotype minnesota Re 595 (Re mutant), Ready Made Solution

1 mg/mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

description

(Ready Made Solution)

Quality Level

form

liquid

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

concentration

1 mg/mL

color

faintly yellow

Mode of action

enzyme | interferes

shipped in

ambient

storage temp.

2-8°C

General description

Lipid A is one of the components of the lipopolysaccharide molecule of Gram-negative bacteria. It is responsible for the toxicity of these bacteria. It has two glucosamine units with a β (1-->6) glycosidic linkage.

Packaging

2ML

Preparation Note

1 mg/mL in aqueous solution containing 1 μL/mL triethylamine

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jolanta Lodowska et al.
Postepy higieny i medycyny doswiadczalnej (Online), 61, 106-121 (2007-03-21)
The majority of Gram-negative bacteria are pathogenic to humans and animals. Lipopolysaccharide (LPS) is the most biologically active component of these microorganisms. This compound is also called endotoxin to emphasize its negative impact on a macroorganism. Lipid A, one of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service