Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAE0203

Sigma-Aldrich

Anti-V5 Magnetic Beads

Magnetic agarose, Suspension

Synonym(s):

Anti-V5 Tag antibody produced in mouse, V5 Antibody, V5 Tag Antibody, V5 epitope Tag

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.56

conjugate

magnetic beads

Quality Level

form

suspension

shipped in

ambient

storage temp.

2-8°C

General description

The Anti-V5 Magnetic Beads are prepared using an affinity-purified anti-V5 antibody produced in mouse conjugated to cyanogen bromide-activated magnetic beads at a protein-to-bead ratio of approximately 2 mg/mL. The Anti-V5 antibody specifically recognizes an epitope located on V5-tagged fusion proteins. It offers specific reactivity with both N-terminal and C-terminal V5-tagged fusion proteins and is suitable for immunoprecipitation or immunoaffinity purification. These magnetic beads can be used for purifying expressed V5-tagged fusion proteins from bacterial lysates, mammalian lysates, or transfected cells and to study protein topology, characterize, and identify new proteins, protein complexes, and associated proteins.

Application

Anti-V5 magnetic beads have been used for the purification of V5-tagged proteins using the process of immunoprecipitation.

Features and Benefits

  • Beads are prepared with an affinity-purified anti-V5 antibody.
  • Monoclonal Anti-V5 antibody is produced by fusing mouse myeloma cells and splenocytes from a BALB/c mouse.
  • Protein-to-bead ratio is approximately 2 mg/mL.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitochondrial amidoxime-reducing component 1 p.Ala165Thr increases protein degradation mediated by the proteasome
Tanmoy Dutta et al.
Liver International : Official Journal of the International Association For the Study of the Liver (2024)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service