Skip to Content
MilliporeSigma

Skip To

SAE0010

EC-Oxyrase®

Synonym(s):

EC-Oxyrase®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.54
UNSPSC Code:
12352204
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

shipped in

dry ice

storage temp.

−20°C

General description

The Oxyrase Enzyme System is obtained from the cytoplasmic membrane of E, coli

Application

Oxyrase is an enzyme system that offers a way to remove dissolved oxygen from liquid, gas or semisolid products. Oxyrase utilizes the same mechanics of respiration employed by all living beings. It selectively captures oxygen and with a substrate converts it to water. Oxyrase is equivalent to a “strong” reducing agent, but unlike chemical antioxidants, Oxyrase, the substrates and reactants are all-natural. Oxyrase is designed to work efficiently over a wide range of pH and temperature
Various applications of Oxyrase include the preparation of anaerobic media for culture of anaerobic microorganisms, and the protection of oxygen-sensitive materials. In studies of biological processes, Oxyrase can help to enhance the intensity and longevity of activated fluorescent dye which are at risk of oxygen quenching.
EC-Oxyrase® has been used as a component of imaging buffer for stochastic optical reconstruction microscopy (STORM) imaging.

Biochem/physiol Actions

EC-Oxyrase is active over a wide temperature range. It can be kept at 40 °C for hours without substantial loss of activity. It is heat inactivated above 55°C.
EC-Oxyrase operates over a wide pH range of 6.8 to 9.4. The optimum pH is about 8.0. As the pH moves away from optimum, activity decreases. The lower activity level can be compensated for by increasing time to complete oxygen removal or by increasing EC-Oxyrase concentration.
EC-Oxyrase requires substrate to be active. Levels at 10 mM to 20 mM are adequate. Substrates for EC Oxyrase include lactic acid, succinic acid, formic acid or their salts, and alpha-glycerol phosphate inaddition to oxygen. EC Oxyrase also contains L-sodium lactate at 100 mM to stabilize the Oxyrase during freeze-thaw cycles. This amount of substrate is diluted to extinction upon use.

The exact volume of EC Oxyrase and substrates needed to reduce oxygen in a given system are determined by a number of parameters: including pH, temperature, kinds and amounts of substrates present, surface to depth ratio of the container, and headspace volume. Some experimentation may be necessary; a suggested use level is a 1:100 dilution of the product.

Analysis Note

EC Oxyrase contains a penicillin binding protein that may interfere with penicillin and some related antibiotics.

Legal Information

Oxyrase is a registered trademark of Oxyrase, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAE0013C5499X4376
shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

300


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters