Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB5200294

Sigma-Aldrich

Monoclonal Anti-Mmp9 antibody produced in mouse

clone S51-82, purified immunoglobulin

Synonym(s):

Anti-82kDa matrix metalloproteinase-9, Anti-CLG4B, Anti-GELB, Anti-MANDP2, Anti-MMP-9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S51-82, monoclonal

form

buffered aqueous solution

mol wt

antigen predicted mol wt 92 & 82 kDa

species reactivity

mouse, human, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

rat ... Mmp9(81687)

Specificity

Detecs ~92kDa and ~82kDa (pro and active forms)

Immunogen

Fusion protein amino acids 1-708 (full length) of rat MMP9

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS pH7.4, 50% glycerol, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Yin et al.
Frontiers in pharmacology, 13, 913098-913098 (2022-08-30)
ShengFu oil is a compounded Chinese medicinal prescription, and provides antibacterial, anti-inflammatory, and analgesic effects, favoring burn wound repair. In this study, we aimed at investigating the effects of topical applications of ShengFu oil and its active ingredients in oral
Jih-Hua Wei et al.
International journal of molecular sciences, 24(6) (2023-03-30)
Bariatric surgery reduces body weight, enhances metabolic and diabetic control, and improves outcomes on obesity-related comorbidities. However, the mechanisms mediating this protection against cardiovascular diseases remain unclear. We investigated the effect of sleeve gastrectomy (SG) on vascular protection in response
Zhi Teng et al.
Oncology reports, 43(2), 503-515 (2020-01-03)
Acute lymphoblastic leukemia (ALL) is a malignant hematological disease. Tanshinone IIA (Tan IIA) has antitumor activity in vitro and in vivo. The aim of the present study was to investigate the effects of Tan IIA in combination with imatinib (IM) on the proliferation, apoptosis, migration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service