Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5200013

Sigma-Aldrich

Monoclonal Anti-AHSA1 antibody produced in mouse

clone 25F2.D9, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-Aha1, Anti-Ahsa1, Anti-C14orf3, Anti-HspC322, Anti-p38

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

25F2.D9, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 38 kDa

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Ahsa1(217737)

Specificity

Detects ~38 kDa. Can run up to 45 kDa on SDS Page.

Immunogen

Mouse Aha1

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in 0.02 M potassium phosphate, 0.15 M sodium chloride, pH 7.2, and 0.09% sodium azide

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service