Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB2101084

Sigma-Aldrich

Anti-HSD11B1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-11-β-HSD1, Anti-11-DH, Anti-HDL, Anti-HSD11, Anti-Hydroxysteroid (11-β) dehydrogenase 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

32 kDa

species reactivity

human, mouse, rat, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSD11B1(3290)

General description

Hydroxysteroid 11-beta dehydrogenase 1 (HSD11B1) short-chain dehydrogenase/reductase superfamily and anchors on the endoplasmic reticulum (ER) membrane. It comprises a hydrophobic N terminus and a catalytic domain. The HSD11B1 gene is mapped to human chromosome location 1q32.2.

Immunogen

Synthetic peptide directed towards the N terminal region of human HSD11B1

Biochem/physiol Actions

Mutation in the hydroxysteroid 11-β dehydrogenase 1 (HSD11B1) gene is implicated in cortisone reductase deficiency.

Sequence

Synthetic peptide located within the following region: QKVVSHCLELGAASAHYIAGTMEDMTFAEQFVAQAGKLMGGLDMLILNHI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole Draper et al.
Nature genetics, 34(4), 434-439 (2003-07-15)
In cortisone reductase deficiency (CRD), activation of cortisone to cortisol does not occur, resulting in adrenocorticotropin-mediated androgen excess and a phenotype resembling polycystic ovary syndrome (PCOS; refs. 1,2). This suggests a defect in the gene HSD11B1 encoding 11beta-hydroxysteroid dehydrogenase type
Jeremy W Tomlinson et al.
Endocrine reviews, 25(5), 831-866 (2004-10-07)
11beta-hydroxysteroid dehydrogenase type 1 (11beta-HSD1) interconverts inactive cortisone and active cortisol. Although bidirectional, in vivo it is believed to function as a reductase generating active glucocorticoid at a prereceptor level, enhancing glucocorticoid receptor activation. In this review, we discuss both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service