Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1406114

Sigma-Aldrich

Anti-MDM4 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp781B1423, HDMX, MDMX, MGC132766, MRP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~54.9 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MDM4(4194)

General description

Mouse double minute 4 (MDM4), also known as MDMX, is encoded by the gene mapped to human chromosome 1q32.1. The encoded protein is a structural homolog of MDM2. It belongs to the MDM family.
The human MDM4 gene, which plays a role in apoptosis, encodes a 490-amino acid protein containing a RING finger domain and a putative nuclear localization signal. The MDM4 putative nuclear localization signal, which all Mdm proteins contain, is located in the C-terminal region of the protein. The mRNA is expressed at a high level in thymus and at lower levels in all other tissues tested. MDM4 protein produced by in vitro translation interacts with p53 via a binding domain located in the N-terminal region of the MDM4 protein. MDM4 shows significant structural similarity to p53-binding protein MDM2. Two transcript variants, one protein-coding and the other likely not to be protein-coding, have been found for this gene. (provided by RefSeq)

Immunogen

MDM4 (NP_002384.2, 1 a.a. ~ 490 a.a) full-length human protein.

Sequence
MTSFSTSAQCSTSDSACRISPGQINQVRPKLPLLKILHAAGAQGEMFTVKEVMHYLGQYIMVKQLYDQQEQHMVYCGGDLLGELLGRQSFSVKDPSPLYDMLRKNLVTLATATTDAAQTLALAQDHSMDIPSQDQLKQSAEESSTSRKRTTEDDIPTLPTSEHKCIHSREDEDLIENLAQDETSRLDLGFEEWDVAGLPWWFLGNLRSNYTPRSNGSTDLQTNQDVGTAIVSDTTDDLWFLNESVSEQLGVGIKVEAADTEQTSEEVGKVSDKKVIEVGKNDDLEDSKSLSDDTDVEVTSEDEWQCTECKKFNSPSKRYCFRCWALRKDWYSDCSKLTHSLSTSDITAIPEKENEGNDVPDCRRTISAPVVRPKDAYIKKENSKLFDPCNSVEFLDLAHSSESQETISSMGEQLDNLSEQRTDTENMEDCQNLLKPCSLCEKRPRDGNIIHGRTGHLVTCFHCARRLKKAGASCPICKKEIQLVIKVFIA

Biochem/physiol Actions

Mouse double minute 4 (MDM4)/ MDMX acts as an essential regulator of the tumor suppressor Tp53. Overexpression of the gene has been observed in patients with hepatocellular carcinoma (HCC). Polymorphism in the gene increases the risk of susceptibility to gastric cancer (GCa). In addition, upregulated expression of the gene is also involved in the etiology of various cancers, such as invasive breast carcinoma liver hepatocellular carcinoma, retinoblastomas and skin cutaneous melanomas.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Etiology?dependent molecular mechanisms in human hepatocarcinogenesis.
Schlaeger C, et al.
Hepatology, 47(2), 511-520 (2008)
MDM4 genetic variants and risk of gastric cancer in an eastern chinese population
Wang MY, et al.
Oncotarget, 8(12), 19547-19547 (2017)
MDM4 actively restrains cytoplasmic mTORC1 by sensing nutrient availability
Mancini F, et al.
Molecular Cancer, 16(1), 55-55 (2017)
The associations between MDM4 gene polymorphisms and cancer risk.
Wang MJ, et al.
Oncotarget, 7(34), 55611-55611 (2016)
A PRISMA-compliant meta-analysis of MDM4 genetic variants and cancer susceptibility
Zhai Y, et al.
Oncotarget, 7(45), 73935-73935 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service