Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1100315

Sigma-Aldrich

Anti-RS1 (66-78) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-RS, Anti-Retinoschisin 1, Anti-XLRS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~26 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RS1(6247)

Related Categories

General description

Retinoschisin 1 (RS1) is a localized in the retina and pineal. Its is encoded by the X-linked retinoschisis (XLRS) gene and is 24 kDa. It consists of a signal sequence, RS1 domain, discoidin domain and a C-terminal domain. It is secreted by the retinal neurons as a homo-octameric complex.

Immunogen

synthetic peptide corresponding to amino acids 66-78 of human RS1

Application

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

Retinoschisin 1 (RS1) plays a role in binding of β-2-laminin, αβ-crystallin, phospholipid, galactose and Na/K ATPase−SARM1 (Sterile alpha and TIR motif-containing protein 1) complex. Mutations in the RS1 gene leads to a condition called as X-linked juvenile retinoschisis which leads to macular degradation in males and deterioration of vision.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular pathology of X linked retinoschisis: mutations interfere with retinoschisin secretion and oligomerisation
Wang T, et al.
The British Journal of Ophthalmology, 90(1), 81-86 (2006)
Inactivation of the murine X-linked juvenile retinoschisis gene, Rs1h, suggests a role of retinoschisin in retinal cell layer organization and synaptic structure
Weber B H F, et al.
Proceedings of the National Academy of Sciences of the USA, 99(9), 6222-6227 (2002)
Intravitreal delivery of AAV8 retinoschisin results in cell type-specific gene expression and retinal rescue in the Rs1-KO mouse
Park T K, et al.
Gene Therapy, 16(7), 916-916 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service