Documents
S9027
Slides, microscope
opaque (white), size 25 mm × 75 mm
About This Item
UNSPSC Code:
41122600
NACRES:
MA.01
material
glass
packaging
case of 20
box of 72 slides
size
25 mm × 75 mm
binding type
non-treated surface
General description
Sealed in a moisture absorbing desiccant pack which can function as a drying stand and a storage box. Slides are 1 mm thick.
Our microscope slides are made of high-quality white glass with low iron content, no green tint with fine texture and transparency; these slides are also corrosion-resistant. The edges are ground smooth to help eliminate sharp cutting surfaces and safe handling. They are packed in a special thermoform plastic box to minimize exposure to moisture and dust (each slide is precleaned).
Conform to Federal specification NNN-S-450.
Visit the Document Library
Customers Also Viewed
Slide 1 of 1
1 of 1
Xiao Yan et al.
ACS nano, 14(10), 12796-12809 (2020-10-15)
Droplet transport on, and shedding from, surfaces is ubiquitous in nature and is a key phenomenon governing applications including biofluidics, self-cleaning, anti-icing, water harvesting, and electronics thermal management. Conventional methods to achieve spontaneous droplet shedding enabled by surface-droplet interactions suffer
Related Content
Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.
Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.
Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.
Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.