Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0208

Sigma-Aldrich

Obestatin EIA Kit

for serum, plasma, culture supernatant and cell lysates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

species reactivity

human, rat, mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

input

sample type culture supernatant(s)
sample type cell lysate
sample type plasma
sample type serum

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 0.1 pg/mL
standard curve range: 0.1–1000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GHRL(51738)

General description

The Obestatin Enzyme Immunoassay (EIA) Kit is an in vitro quantitative assay for detecting Obestatin peptide based on the principle of Competitive Enzyme Immunoassay.

Immunogen

Obestatin, synthetic peptide

Kit Components Also Available Separately

Product No.
Description
SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Evaluation of Acylated Ghrelin and Obestatin Levels in Childhood Obesity and Their Association with Insulin Resistance, Metabolic Syndrome, and Oxidative Stress.
Razzaghy-Azar M
Journal of clinical medicine, 5, E61-E61 (2016)
Yongfeng Yang et al.
Behavioral and brain functions : BBF, 8, 11-11 (2012-03-01)
Ghrelin (GHRL) is a pivotal peptide regulator of food intake, energy balance, and body mass. Weight gain (WG) is a common side effect of the atypical antipsychotics (AAPs) used to treat schizophrenia (SZ). Ghrelin polymorphisms have been associated with pathogenic
Philippe Zizzari et al.
Endocrinology, 148(4), 1648-1653 (2007-01-06)
Administration of ghrelin, an endogenous ligand for the GH secretagogue receptor 1a (GHSR 1a), induces potent stimulating effects on GH secretion and food intake. However, more than 7 yr after its discovery, the role of endogenous ghrelin remains elusive. Recently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service